Skip to Content
MilliporeSigma

49596

Multielement standard solution 3 for ICP

TraceCERT®, in 5% nitric acid

Synonym(s):

ICP multielement standard solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

quality

in 5% nitric acid

composition

Ca, 2000 mg/L , K, 200 mg/L , Mg, 400 mg/L , Na, 1000 mg/L

concentration

in 5% HNO3

technique(s)

ICP: suitable

application(s)

industrial qc
pharmaceutical

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Search & download your certificate here.

Application

Multielement standard solution 3 for ICP can be used as a calibration standard in elemental analysis for inductively coupled plasma optical emission spectrometry (ICP-OES) method.

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

supp_hazards

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library