Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.09200

Millipore

Mains charger

for use with MAS-100 NT®, with Lemo plug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

plastic hardware

Agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with MAS-100 NT® Ex
for use with MAS-100 NT®

storage temp.

room temp

General description

Mains charger is an additional accessory for MAS-100 NT® air sampler. The MAS-100 NT® is an industry standard air sampler, designed to monitor air microbiology in cleanrooms and hygienically ultra-sensitive areas. Its high recovery rate is based on the Andersen impaction principle. An integrated airflow sensor compensates for differences in pressure and temperature in the environment. The Mains charger for MAS-100 NT® has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ±5% (2.7 A). The cable length varies depending upon the country.

Application

Mains charger is an additional accessory for MAS-100 NT® air sampler that is used for microbial monitoring of ambient air in controlled environments up to ISO 5 cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Linkage

Replaces: 4.86729.0001

Legal Information

MAS-100 NT is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service