Skip to Content
MilliporeSigma

1.09784

MAS-100 Iso MH® Power Supply

including cord for EU, GB, US, JP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

aluminum hardware

agency

non-compendial

feature

electronic

packaging

pkg of 1 kit

application(s)

air monitoring
pharmaceutical

compatibility

for use with MAS-100 Iso MH®

shipped in

ambient

storage temp.

no temp limit

General description

MAS-100 ISO MH® Power Supply is an additional accessory designed to be specifically used for MAS-100 ISO MH® air sampler. The MAS-100 ISO MH® air sampler has been specially developed for microbial monitoring of ambient air is isolators, RABS and high-grade cleanrooms. The Power supply has an input voltage of 100 to 240 V AC (47 to 63 Hz, 1.5 to 0.7 A) and output voltage of 24 V DC ±5% (2.7 A). The power supply also has a cable conductor cross-chapter of 18AWG and Switchcraft S761KS12 with a center pin of 2.0 mm connector.

Application

MAS-100 ISO MH® Power Supply is an additional accessory. MAS-100 ISO MH® air sampling devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 ISO MH is a registered trademark of MBV AG




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library