Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09194

Millipore

MAS-100 NT® Ex

suitable for air monitoring in explosion hazard areas

Synonym(s):

MAS-100 NT® Ex microbial air monitoring system, microbial air monitoring systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103403
NACRES:
NA.74

material

aluminum hardware

Agency

according to ISO 14698

feature

portable ( )

packaging

pkg of 1 kit

manufacturer/tradename

MAS-100 NT® Ex

weight

2.4 kg (including perforated lid)

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

90 mm Petri dishes

General description

The MAS-100 NT® Ex microbial air monitoring systems are high performance, portable, battery operated instrument which are based on the principle of the Andersen air sampler. These compact yet sophisticated devices are the preferred choice for those demanding the highest quality in microbial air monitoring. An ambient amount of air is aspirated through a 300-hole perforated lid and impacted onto the surface of growth media in standard 90-100 mm Petri dishes or 55-60 mm contact plates. Microorganisms are impacted onto the agar and, after an appropriate incubation period, the colonies are counted. The system measures the airflow and regulates it to a constant value of 100 standard liters/min A Standard Liter Per Minute (SLPM) is adjusted to a specific environmental temperature and pressure, this ensure the amount of air sampled is always the same and independent of environmental conditions. These systems have the highest airflow accuracy of ±2.5%, compared to others that can be as high as ±10%. The integrated flow sensor allows the user to freely interchange the perforated lids without affecting the accuracy or the calibration of the unit. The MAS-100 NT® Ex is powered by a Lithium ion rechargeable battery with an intelligent charging program that assures long battery life without routine discharging.

Application

The MAS-100 NT® Ex air sampler is used for microbial monitoring of ambient air in explosion hazard areas. Microbial air sampling devices are extensively used in Pharmaceutical and Personal care industries as well as Food, Beverage and Environmental applications.

Components

Includes hard carrying case, battery, 300-hole perforated lid, dust cover, main charger, USB cable, 3 mm allen key, USB stick with all documentation

Legal Information

MAS-100 NT is a registered trademark of MBV AG

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Food microbiology testing is a key part of the food safety and quality management. Learn more about the wide range of tests covering pathogens and spoilage organisms.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service