Skip to Content
MilliporeSigma
All Photos(4)

Documents

28118-U

Supelco

Escort Elf Twin Port Sampling Pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

product name

Escort Elf Twin Port Sampler,

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

General description

This pump attachment is designed for low flow industrial hygiene sampling, such as gas or vapor monitoring, using sorbent tubes. Two needle valves provide independent flow control for simultaneous collection on two tubes but, by closing the flow to one valve, a single tube can be used. The sampler is compatible with any personal sampling pump capable of a 1.5 Lpm flow rate at a load of 25 inches of water. Total flow cannot exceed 500 mL/min. Each sampler comes with two tube protectors, one for small tubes (<2 in. long) and one for large tubes (<41/2 in. long), and tubing required to connect the sampler to the sampling pump.

Features and Benefits

Features:

  • Valved mechanism allows flow control down to .001 L/min
  • Control the flow rate of each port independently
  • Includes tube covers to fit both large and small tube sizes

Legal Information

Escort is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Learn more about how the thermal desorption technique is applied for terpene identification, profiling, odor measurement, industrial hygiene, and environmental emissions.

Protocols

Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service