Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.07256

Millipore

L-Phenylalanine

for biochemistry

Synonym(s):

L-Phenylalanine, Phe, 2-Amino-3-phenylpropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO2
CAS Number:
Molecular Weight:
165.19
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-568-1
NACRES:
NA.21

vapor pressure

<1 Pa ( 25 °C)

Quality Level

form

solid

pH

5.4-6.0 (20 °C, 10 g/L in H2O)

mp

275-283 °C (decomposition)

solubility

27 g/L

density

1.34 g/cm3 at 20 °C

bulk density

580 kg/m3

storage temp.

2-30°C

InChI

1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)

InChI key

COLNVLDHVKWLRT-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): 99.0 - 101.0 %
Identity (IR-spectrum): passes test
Identity (specific rotation): passes test
Appearance: white to almost white, crystalline powder
Appearance of solution (50 g/l, hydrochloric acid 1 mol/l): clear and colorless
Spec. rotation (α 20/D, 20 g/l, water, calc. on dried substance): -33,0 to -35,5
Chloride (Cl): ≤ 200 ppm
Sulfate (SO₄): ≤ 300 ppm
Al (Aluminium): ≤ 5 ppm
As (Arsenic): ≤ 5 ppm
Ca (Calcium): ≤ 10 ppm
Co (Cobalt): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
K (Potassium): ≤ 5 ppm
Mg (Magnesium): ≤ 5 ppm
Na (Sodium): ≤ 50 ppm
Zn (Zinc): ≤ 5 ppm
Ninhydrin-positive substances (LC)(any ninhydrin-positive impurity): ≤ 0.2 %
Ninhydrin-positive substances (LC) (ammonium (570 nm)): ≤ 0.02 %
Ninhydrin-positive substances (LC)(total impurities): ≤ 0.5 %
Loss on drying (105 °C; 3 h): ≤ 0.3 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service