Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.06525

Supelco

Sodium iodate

for analysis EMSURE®

Synonym(s):

Sodium iodate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
INaO3
CAS Number:
Molecular Weight:
197.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-672-5
NACRES:
NA.21
form:
solid
solubility:
81 g/L

Quality Level

product line

EMSURE®

Assay

≥99.5% (iodometric)

form

solid

pH

5.8 (25 °C, 90 g/L in H2O)

solubility

81 g/L

density

4.28 g/cm3 at 20 °C

bulk density

1150 kg/m3

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤0.02%
iodide (I-): ≤0.002%
sulfate (SO42-): ≤0.005%

cation traces

Cd: ≤0.00005%
Cu: ≤0.0001%
Fe: ≤0.001%
K: ≤0.02%
Mn: ≤0.00005%
N: ≤0.002%
Pb: ≤0.0002%
Zn: ≤0.0001%

storage temp.

2-30°C

SMILES string

[Na+].[I](=O)(=O)[O-]

InChI

1S/HIO3.Na/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

WTCBONOLBHEDIL-UHFFFAOYSA-M

Application


  • Comparative Analysis of Molecular Landscape in Mouse Models and Patients Reveals Conserved Inflammation Pathways in Age-Related Macular Degeneration.: This study highlights the use of sodium iodate to induce retinal degeneration in mouse models to explore conserved inflammation pathways relevant to age-related macular degeneration. This model provides insights into potential therapeutic targets for inflammatory diseases of the retina (Liu et al., 2024).

  • The BET PROTAC inhibitor dBET6 protects against retinal degeneration and inhibits the cGAS-STING in response to light damage.: Sodium iodate was utilized to model retinal degeneration, showing the effectiveness of the BET PROTAC inhibitor dBET6 in protecting against retinal damage and modulating inflammatory responses, potentially offering new treatment avenues for retinal diseases (Zhu et al., 2023).

  • Sodium Iodate-Induced Degeneration Results in Local Complement Changes and Inflammatory Processes in Murine Retina.: This research delves into the specific inflammatory processes and complement system changes following sodium iodate-induced retinal degeneration in mice, providing a detailed molecular understanding of retinal inflammatory responses (Enzbrenner et al., 2021).

  • IL-33trap-mediated IL-33 neutralization does not exacerbate choroidal neovascularization, but fails to protect against retinal degeneration in a dry age-related macular degeneration model.: The study used sodium iodate to induce retinal degeneration and test IL-33 neutralization′s effectiveness in preventing retinal damage, contributing to the understanding of inflammatory pathways in retinal diseases (Van Hove et al., 2021).

  • An Acute Injury Model for the Phenotypic Characteristics of Geographic Atrophy.: Utilizing sodium iodate to create an acute retinal injury model, this study mimics the phenotypic characteristics of geographic atrophy, providing a valuable tool for studying this condition and testing potential treatments (Bhutto et al., 2018).

Analysis Note

Assay (iodometric): ≥ 99.5 %
pH-value (5 %; water): 5 - 8
Chloride, Chlorate, Bromide, Bromate (as Cl): ≤ 0.02 %
Iodide (I): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.005 %
Total nitrogen (N): ≤ 0.002 %
Cd (Cadmium): ≤ 0.00005 %
Cu (Copper): ≤ 0.0001 %
Fe (Iron): ≤ 0.001 %
K (Potassium): ≤ 0.02 %
Mn (Manganese): ≤ 0.00005 %
Pb (Lead): ≤ 0.0002 %
Zn (Zinc): ≤ 0.0001 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service