Skip to Content
MilliporeSigma

1.01047

Aluminium potassium sulfate dodecahydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Aluminium potassium sulfate dodecahydrate, Potassium aluminium sulfate, Alum potassium, Potassium alum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
KAl(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
474.39
UNSPSC Code:
12352302
EC Index Number:
233-141-3
NACRES:
NA.21
MDL number:
Assay:
99.0-100.5% (complexometric)
Grade:
ACS reagent
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

agency

reag. Ph. Eur.

product line

EMSURE®

assay

99.0-100.5% (complexometric)

form

solid

impurities

≤0.005% Insoluble matter

pH

3.0-3.5 (20 °C, 100 g/L in H2O)

mp

92.5 °C

density

1.75 g/cm3 at 20 °C

bulk density

900 kg/m3

anion traces

chloride (Cl-): ≤0.0005%

cation traces

Cd: ≤0.0005%, Cu: ≤0.0005%, Fe: ≤0.0005%, NH4+: ≤0.005%, Na: ≤0.005%, Pb: ≤0.0005%, heavy metals: ≤0.001% (ACS)

storage temp.

2-30°C

SMILES string

[K+].[Al]1(O[S](=O)(=O)O1)O[S](=O)(=O)[O-].O.O.O.O.O.O.O.O.O.O.O.O

InChI

1S/Al.K.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;;2*(H2,1,2,3,4);12*1H2/q+3;+1;;;;;;;;;;;;;;/p-4

InChI key

GNHOJBNSNUXZQA-UHFFFAOYSA-J

Application


  • Extraction and characterization of lignin from oil palm biomass via ionic liquid dissolution and non-toxic aluminium potassium sulfate dodecahydrate precipitation processes.: This study explores an environmentally benign method for extracting lignin from oil palm biomass using ionic liquids, followed by the precipitation of lignin using non-toxic aluminium potassium sulfate dodecahydrate. The process is notable for its eco-friendliness and efficiency in lignin recovery, highlighting a potential application in bio-refining technologies (Mohtar et al., 2015).

  • Reduction of house-dust mite allergen concentrations in carpets by aluminium potassium sulfate dodecahydrate (alum).: This research investigates the effectiveness of aluminium potassium sulfate dodecahydrate (alum) in reducing allergen concentrations in household carpets. The study demonstrates alum′s potential as a simple and cost-effective method to significantly decrease allergen levels, thus contributing to improved indoor air quality and reducing allergen-related health issues (Sevki et al., 2006).

Analysis Note

Assay (complexometric): 99.0 - 100.5 %


Identity: passes test


Appearance of solution: passes test


Insoluble matter: ≤ 0.005 %


pH-value (10 %; in water): 3.0 - 3.5


Chloride (Cl): ≤ 0.0005 %


Heavy metals ( ACS): ≤ 0.001 %


Cd (Cadmium): ≤ 0.0005 %


Cu (Copper): ≤ 0.0005 %


Fe (Iron): ≤ 0.0005 %


Na (Sodium): ≤ 0.005 %


NH₄ (Ammonium): ≤ 0.005 %


Pb (Lead): ≤ 0.0005 %


Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?


Storage Class

13 - Non Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library