Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

P5958

Sigma-Aldrich

Potassium hydroxide

BioXtra, ≥85% KOH basis

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
pellets
solubility:
H2O: 1 M, clear, colorless

grade

for analytical purposes

vapor pressure

1 mmHg ( 719 °C)

product line

BioXtra

Assay

≥85% KOH basis

form

pellets

impurities

≤0.0005% Phosphorus (P)
≤0.1% Insoluble matter
≤2.0% K2CO3

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.003%

cation traces

Al: ≤0.0005%
Ca: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Na: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide is an inorganic compound commonly known as caustic potash. It is a strong alkali, readily reacts with acids. It can be used in various reactions such as nucleophilic substitution, hydrolysis, and addition reactions. Sometimes it can also be used as a catalyst for aldol-type reactions.

Application

Potassium hydroxide can be used to adjust the pH of the solution.
It can also be used:
  • As a catalyst in the alkynylation of heteroaromatic N-oxides to synthesize corresponding C2-alkynylheterocycles.
  • As a superbase medium with dimethyl sulfoxide (KOH/DMSO medium) for the synthesis of cross-coupling products under transition metal-free conditions from reactions between aryl halides and various sulfur, oxygen and nitrogen-based nucleophiles.
  • In the synthesis and thermodynamic study of Na-based zeolites.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuqian Luo et al.
BMC immunology, 15, 586-586 (2014-12-21)
Highly purified nuclear protein is required when using an electrophoretic mobility shift assay (EMSA) to study transcription factors, e.g. nuclear factor-κB (NF-κB), a major transcription factor that regulates both innate and adaptive immune responses following infection. Although many protocols have
Iulia I Nita et al.
PloS one, 7(10), e46649-e46649 (2012-10-12)
Mitochondria mediate dual metabolic and Ca(2+) shuttling activities. While the former is required for Ca(2+) signalling linked to insulin secretion, the role of the latter in β cell function has not been well understood, primarily because the molecular identity of
Jae Myoung Suh et al.
Nature, 513(7518), 436-439 (2014-07-22)
Fibroblast growth factor 1 (FGF1) is an autocrine/paracrine regulator whose binding to heparan sulphate proteoglycans effectively precludes its circulation. Although FGF1 is known as a mitogenic factor, FGF1 knockout mice develop insulin resistance when stressed by a high-fat diet, suggesting a
Marija Smokvarska et al.
Current biology : CB, 30(23), 4654-4664 (2020-10-10)
In the course of their growth and development, plants have to constantly perceive and react to their environment. This is achieved in cells by the coordination of complex combinatorial signaling networks. However, how signal integration and specificity are achieved in
Guilhem Reyt et al.
Current biology : CB, 30(20), 4103-4111 (2020-08-29)
Casparian strips (CSs) are cell wall modifications of vascular plants restricting extracellular free diffusion into and out of the vascular system [1]. This barrier plays a critical role in controlling the acquisition of nutrients and water necessary for normal plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service