Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.04070

Millipore

Gelatine

suitable for microbiology

Synonym(s):

Gelatine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
EC Index Number:
232-554-6
NACRES:
NA.74

biological source

animal

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)

pH

3.8-7.6 (25 °C, 10 g/L in H2O)

application(s)

microbiology

storage temp.

15-25°C

Analysis Note

Identity (A: coloring): passes test
Identity (B: sediment): passes test
Identity (C: Falling out): passes test
Identity (NIR): passes test
pH-value (1 %; water): 5 - 6
Sulfur dioxide (SO₂): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.005 %
Peroxide (as H₂O₂): ≤ 0.01 %
Sulfated ash (800 °C): ≤ 2 %
Loss on Drying (105°C): ≤ 15 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service