Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.08635

Sigma-Aldrich

Kaiser′s Glycerol Gelatine

phenol-free for microscopy

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.21

Quality Level

IVD

for in vitro diagnostic use

color

light yellow

density

1.117-1.120 g/cm3 at 40 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Kaiser′s glycerol gelatin - phenol-free for microscopy, is an aqueous, phenol-free mounting medium consisting of gelatine and glycerol, and is used primarily in the mounting of stained histology specimens (e. g. HEMATOGNOST Fe - staining kit (Product number 1.12084). The pH of the product lies in the neutral range. Prior to use, the mounting medium is melted in the heating cabinet at approx. 40 °C and is dropped onto the specimen from the practical dropping bottle. The preservative phenol has been replaced with a proprietary compound that is more environmentally friendly. This affords the unique advantage of eliminating phenol fumes when the medium is heated before use. CE-certified, IVD aqueous mounting media developed specifically for use in histological applications is essential in the support of formal quality assurance programs to monitor staining performance in pathology laboratories. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Aqueous mounting medium

Analysis Note

Suitability for microscopypasses test

Legal Information

HEMATOGNOST FE is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service