Skip to Content
MilliporeSigma
All Photos(1)

Documents

73865

Millipore

Gelatin

from Porcine skin, pkg of 500 g, suitable for microbiology

Synonym(s):

Gelatin from porcine skin, Collagen hydrolysate, Collagen peptides, Gelatin hydrolysate, Hydrolyzed collagen, Hydrolyzed gelatin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74

biological source

Porcine skin

Quality Level

form

powder

quality

controlled endotoxin level
medium gel strength

shelf life

36 mo. (Expiry date on the label)

packaging

pkg of 500 g

impurities

≤2.0% sulfated ash
≤5 EU/mg endotoxins

loss

≤15% loss on drying

color

faint beige to light beige
faint brown to light brown
white to light yellow

pH

4.0-6.0

gel strength

150-210 g Bloom

solubility

H2O: 0.67 g/10 mL at 50 °C (colorless to very light yellow, clear to very hazy)

anion traces

chloride (Cl-): ≤3000 mg/kg

cation traces

Ca: ≤2000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤10 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤500 mg/kg
Mg: ≤500 mg/kg
Mn: ≤5 mg/kg
Na: ≤5000 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

application(s)

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Gelatin is a versatile, high-quality hydrocolloid derived from porcine skin. It is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. noted for its ultrahigh gel strength, which is an important characteristic for various microbiological procedures where a firm gel is required, such as in culture media or other applications where the gel′s structural integrity is crucial.

Application

Gelatin is commonly used as a gelling agent in the preparation of solid and semi-solid culture media. It also aids in the study of aerobic and anaerobic microorganisms by providing a supportive structure for colony formation and development.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service