Skip to Content
MilliporeSigma

1.04017

Glass beads 5 mm

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.51
UNSPSC Code:
41122409
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


storage temp.

no temp limit

Quality Segment

General description

Glass beads can be used to purify, enrich, or separate samples of solids, liquids, or gases. They offer excellent batch consistency to ensure reliable, reproducible results.

Application


  • Design of closed loop for reproduction of gas-dominated hydrate flow: Utilizes 5 mm glass beads in a novel experimental setup to study the flow dynamics of gas hydrates, which is crucial for developing safer and more efficient energy transport systems (Wada et al., 2024).

  • Physicochemical cell disruption of Bacillus sp. for recovery of polyhydroxyalkanoates: This study highlights the role of 5 mm glass beads in the effective disruption of bacterial cells, enhancing the recovery of bioplastics and contributing to sustainable material production (Bhat et al., 2024).

  • Preparation and Characterization of TiO(2)-Coated Hollow Glass Beads for Functionalization of Deproteinized Natural Rubber Latex: Demonstrates the use of 5 mm glass beads coated with TiO2 to improve the mechanical properties and UV resistance of natural rubber, essential for developing advanced materials (Nijpanich et al., 2023).

  • Halotolerant bacterial biofilms for desalination and water treatment: Employs 5 mm glass beads as biofilm carriers to enhance the efficiency of bacterial processes in water purification systems, addressing global challenges in water scarcity (Gauhar et al., 2023).

Analysis Note

Identity: conforms




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library