Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09161

Supelco

Ferroin solution

redox indicator solution, for waste water analysis

Synonym(s):

Ferroin Redox indicator solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Ferroin indicator solution, for waste water analysis

Quality Level

form

liquid

pH

4-7 (20 °C in H2O)

density

1.01 g/cm3 at 20 °C

storage temp.

15-25°C

Analysis Note

Spec. Absorptivity A 1%/1cm (λmax; 0.2 %; buffer pH 4.0): 2.4 - 2.8
pH-value (20 °C): 3.5 - 4.5
Application test: conforms

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Application Note details preparation of peracetic acid standard solution for analytical quality assurance (AQA) in testing laboratories.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service