Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
Ag2O4S
CAS Number:
Molecular Weight:
311.80
UNSPSC Code:
12352302
EC Index Number:
233-653-7
NACRES:
NB.21
MDL number:
grade
ACS reagent
Quality Segment
potency
5000 mg/kg LD50, oral (Rat)
analyte functional class(es)
chemical oxygen demand (COD)
technique(s)
photometry: suitable
pH
5-6 (25 °C, 5 g/L in H2O)
mp
655 °C
solubility
8 g/L
density
5.45 g/cm3 at 20 °C
bulk density
1200 kg/m3
storage temp.
no temp limit
SMILES string
[Ag+].[Ag+].[S](=O)(=O)(O)O
InChI
1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;
InChI key
YPNVIBVEFVRZPJ-UHFFFAOYSA-N
Application
- Lab on a bead with oscillatory centrifugal microfluidics for fast and complete mixing enables fast and accurate biomedical assays: The integration of tetramethylbenzidine in a novel lab-on-a-bead platform demonstrates a significant improvement in the processing speed and reliability of biochemical assays (Williams et al., 2024).
Analysis Note
Assay (argentometric): ≥ 98.5 %
Identity: passes test
Insoluble matter and silver chloride: ≤ 0.02 %
Substances not precipitated by hydrochloric acid (as SO₄): ≤ 0.03 %
Chloride (Cl): ≤ 10 ppm
Nitrate (NO₃): ≤ 10 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Pb (Lead): ≤ 10 ppm
Corresponds to ACS
Identity: passes test
Insoluble matter and silver chloride: ≤ 0.02 %
Substances not precipitated by hydrochloric acid (as SO₄): ≤ 0.03 %
Chloride (Cl): ≤ 10 ppm
Nitrate (NO₃): ≤ 10 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
Pb (Lead): ≤ 10 ppm
Corresponds to ACS
Still not finding the right product?
Explore all of our products under Silver sulfate
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

