Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02052

SAFC

Calcium acetate hydrate

extra pure

Pharma Manufacturing

Synonym(s):

Calcium acetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · x H2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-540-9

Quality Level

form

solid

autoignition temp.

680-730 °C

potency

4280 mg/kg LD50, oral (Rat)

pH

7.2-7.8 (20 °C, 50 g/L in H2O)

solubility

400 g/L

density

1.5 g/cm3 at 20 °C

application(s)

pharma/biopharma processes

storage temp.

2-30°C

Related Categories

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Calcium acetate hydrate is used as a pH modifier and stabilizer in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service