Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.09325

Supelco

Calcium acetate hydrate

[about 94% Ca(CH₃COO)₂] for soil testing

Synonym(s):

Calcium acetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · x H2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-540-9
NACRES:
NA.21

Quality Level

form

powder

autoignition temp.

680-730 °C

potency

4280 mg/kg LD50, oral (Rat)

pH

7.2-7.8 (20 °C, 50 g/L in H2O)

solubility

400 g/L

density

1.5 g/cm3 at 20 °C

bulk density

400 kg/m3

storage temp.

no temp limit

Analysis Note

Assay (complexometrically, Ca(CH₃COO)₂): 93.0 - 95.0 %
Identity (Ca): conforms
Identity (acetate): conforms
Appearance: white to almost white fine powder
Appearance of solution (100 g/l, water): clear and colorless
pH (50 g/l; water): 7.2 - 7.8
Total nitrogen (N): ≤ 0.002 %
Chloride (Cl): ≤ 100 ppm
Sulphate (SO₄): ≤ 1000 ppm
Heavy metals (as Pb): ≤ 10 ppm
Fe (Iron): ≤ 10 ppm
K (Potassium): ≤ 0.02 %
Na (Sodium): ≤ 0.02 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Turbidimetric determination subsequent to extraction with CAL buffer analogous LUFA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service