Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.02035

Supelco

Dragendorff′s reagent spray solution

for thin-layer chromatography

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
NACRES:
NB.21

Quality Level

form

liquid

technique(s)

thin layer chromatography (TLC): suitable

bp

77 °C/1013 hPa

density

0.95 g/cm3 at 20 °C

storage temp.

15-25°C

General description

Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.

Application


  • Metabolite profiling and histochemical localization of alkaloids in Hippeastrum papilio (Ravena) van Scheepen.: This study employed Dragendorff′s reagent for the histochemical localization of alkaloids in the plant species, providing insights into the distribution and concentration of these compounds (Haist et al., 2024).

  • Acetogenins from the Stem of Uvaria rufa and Their Cytotoxic Activity.: This research identified and characterized acetogenins from Uvaria rufa using Dragendorff′s reagent among other methods, highlighting its utility in detecting these potent compounds with potential therapeutic applications (Gurgul et al., 2024).

  • A novel smartphone HPTLC assaying platform versus traditional densitometric method for simultaneous quantification of alfuzosin and solifenacin in their dosage forms as well as monitoring content uniformity and drug residues on the manufacturing equipment.: Incorporates Dragendorff′s reagent in a novel high-performance thin-layer chromatography (HPTLC) method for drug quantification, enhancing the flexibility and accessibility of pharmaceutical analysis (Wadie et al., 2023).

  • Rapid Detection and Determination of Scopolamine in the Leaf Extract of Black Henbane (Hyoscyamus niger L.) Plants Using a Novel Nanosensor.: This study uses Dragendorff′s reagent in a novel nanosensor for the rapid detection of scopolamine, demonstrating its critical role in developing new analytical technologies (Mousavizadeh et al., 2022).

  • Structural and histochemical aspects in leaves of six species of Anemia (Anemiaceae) occurring in rocky outcrops.: Examines the structural and chemical properties of Anemia plants, utilizing Dragendorff′s reagent to reveal the presence and distribution of specific phytochemicals within the plants (Moraes Neto et al., 2022).

Analysis Note

Application test: passes test

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service