MilliporeSigma
58005
Reagent sprayer for TLC plates
volume 250 mL, Bottle-Type
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41115711
NACRES:
NB.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
technique(s)
thin layer chromatography (TLC): suitable
volume
250 mL
General description
The extraction phase or adsorbent phase can be coated on to the solid phase as a thin film. Coating preparation is of three types; dipping, brush painting and spraying. Spraying method was widely used due to its stability and reusability.
Application
Compressed-air sprayers for spraying TLC plates. Sets include a glass-sprayer head and a choice of either bottle or tube with hexfoot stand.
Reagent sprayer may be used in thin-layer chromatography.
Choose from one of the most recent versions:
Certificates of Analysis (COA)