Skip to Content
MilliporeSigma

1.06705

Ninhydrin spray solution

for thin-layer chromatography

Synonym(s):

Ninhydrin Reagent Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.21
UNSPSC Code:
41115711
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

42.4 hPa ( 20 °C)

Quality Segment

form

liquid

autoignition temp.

425 °C (2-Propanol)

expl. lim.

2-13.4 % (v/v) 2-Propanol()

technique(s)

thin layer chromatography (TLC): suitable

bp

82.4 °C/1013 hPa (2-Propanol)

mp

-89.5 °C (2-Propanol)

transition temp

flash point 12 °C (2-Propanol)

storage temp.

15-25°C

General description

Our TLC accessories allow professionals to work more quickly, more efficiently, and more reliably. Thanks to our broad assortment, users will always find what they need.

Analysis Note

Application test: passes test


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

53.6 °F - (2-Propanol)

flash_point_c

12 °C - (2-Propanol)



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library