Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.01812

Supelco

Sulfate Test, photometric

photometric, 0.50-50.0 mg/L (SO42-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Sulfate Test, photometric, 0.50-50.0 mg/L (SO42-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests

specific analyte(s)

sulfate

measuring range

0.50-50.0 mg/L (SO42-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Barium sulfate, turbidimetric)

storage temp.

15-25°C

General description

Method: photometric 0.50 - 50.0 mg/l SO₄²⁻ Spectroquant®

The method is analogous to EPA 375.4, APHA 4500-SO42- E, and ASTM D516-11.

This Spectroquant® Sulfate Reagent Test allows the accurate quantification of the sulfate content in aqueous samples.

Method applied: Sulfate ions react with barium ions to form slightly soluble barium sulfate. The resulting turbidity is measured in the photometer (turbidimetric method).

The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Protocols

Photometric or turbidimetric determination subsequent to water extraction/fusion melting

Photometric or turbidimetric determination subsequent to water extraction/fusion melting

Photometric or turbidimetric determination subsequent to water extraction/fusion melting

Photometric or turbidimetric determination subsequent to water extraction/fusion melting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service