Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.10019

Supelco

Sulfate Test Strips, colorimetric

200-1600 mg/L (SO₄²⁻), MQuant®, for use with MQuant® StripScan App

Synonym(s):

Sulfate chemical test strips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Sulfate Test, colorimetric, 200-1600 mg/L (SO42-), for use with MQuant® StripScan App, MQuant®

description

semi-quantitative

Quality Level

product line

MQuant®

specific analyte(s)

sulfate

measuring range

200-1600 mg/L (SO42-)

compatibility

for use with MQuant® StripScan App

detection method

colorimetric

storage temp.

15-25°C

General description

Method: Colorimetric, with test strips, 200 - 400 - 800 - 1200 - 1600 mg/L (SO42-), MQuant®

The MQuant® Sulfate test strips can be used to give a rapid estimation of the sulfate ion content of drinking and industrial water, as well as of effluent discharged, for instance, from electroplating works and leather goods manufacturers. Sulfate ions react with a red thorin-barium complex, releasing yellow thorin in the process. The sulfate concentration is measured semi-quantitatively by visual comparison of the reaction zones of the test strip with the color rows of a color scale. The test strips are perfect for pre-testing a sample in an economic and quick way to estimate the sulfate concentration and a possible dilution factor prior to using a quantitative method such as a Spectroquant® Sulfate test.

Linkage

Replaces: 10019-1; 10019

Legal Information

MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

MQuant® StripScan app delivers instant, reliable results for various test strips on mobile phones, including pH and hardness.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service