Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.01468

Supelco

Anthrone

GR for analysis ACS,Reag. Ph Eur

Synonym(s):

Anthrone, 9,10-Dihydro-9-oxoanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10O
CAS Number:
Molecular Weight:
194.23
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-994-0
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

mp

151-156 °C

solubility

4.6 mg/L

bulk density

450 kg/m3

storage temp.

15-25°C

InChI

1S/C14H10O/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-8H,9H2

InChI key

RJGDLRCDCYRQOQ-UHFFFAOYSA-N

Analysis Note

Purity (GC): ≥ 98.0 %
Identity (IR-spectrum): conforms
Appearance: Almost white pale yellow to greenish yellow crystal needles or crystalline powder.
Solubility in ethyl acetate (20 g/l): conforms
Appearance of solution (Into Toluol (50 g/l)): conforms
Appearance of solution(Into Sulfuric acid (96 %, 20 g/l)): conforms
Melting point: 153 - 157 °C
Absorbance of reagent solution(425 nm; 0,5 g/l; sulfuric acid 65 %): ≤ 0.75
Absorbance of reagent solution(620 nm; 0,5 g/l; sulfuric acid 65 %): ≤ 0.045
Fe (Iron): ≤ 0.001 %
Sensitivity to carbohydrates(A (620 nm, Glucose 750 µg/25 ml)): ≥ 0.90
Sensitivity to carbohydrates (linearity): conforms
Water (according to Karl Fischer): ≤ 0.5 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service