Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
ACS reagent
Quality Segment
assay
97%
form
powder or crystals
technique(s)
titration: suitable
color
off-white to yellow
mp
154-157 °C (lit.)
solubility
ethyl acetate: 2%, clear, yellow
Amax
≤0.045 at 620 nm in sulfuric acid, ≤0.75 at 425 nm in sulfuric acid
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
O=C1c2ccccc2Cc3ccccc13
InChI
1S/C14H10O/c15-14-12-7-3-1-5-10(12)9-11-6-2-4-8-13(11)14/h1-8H,9H2
InChI key
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
General description
Application
- The anthrone method is a colorimetric assay to detect reducing and nonreducing sugars.
- Anthrone is used in the quantitative determination of streptomycin.
- It has been used to detect carbohydrate containing lipids and to measure sugar content such as sucrose and trehalose in the liquid medium.
1 of 1
This Item | |||
|---|---|---|---|
| description ACS reagent, 97% | description ACS reagent | description ACS reagent | description ACS reagent |
| grade ACS reagent | grade ACS reagent | grade ACS reagent | grade ACS reagent |
| technique(s) titration: suitable | technique(s) titration: suitable | technique(s) titration: suitable | technique(s) titration: suitable |
| color off-white to yellow | color - | color - | color very dark brown to very dark red |
| form powder or crystals | form powder | form powder | form powder or crystals |
| application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing | application(s) diagnostic assay manufacturing |
| solubility ethyl acetate: 2%, clear, yellow | solubility water: 0.77 g/L at 100 °C | solubility ethanol: 0.1% | solubility - |
Still not finding the right product?
Explore all of our products under Anthrone
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




