Skip to Content
MilliporeSigma

1.00045

TOS-MUP Selective Supplement (Mupirocin)

For the preparation of TOS-MUP agar, For the enumeration of Bifidobacteria

Synonym(s):

Selective supplement MUP, MUP supplement, MUP Selective Supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 29981

Quality Segment

form

lyophilized powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

compatibility

for use with TOS-Propionate Agar (Base) (1.00043)

storage temp.

2-8°C

suitability

Bifidobacterium spp.

General description

MUP Selective Supplement contains lithium mupirocin in lyophilized form. Lithium mupirocin selectively inhibits the growth of typical lactic acid bacteria from dairy products, such as Lactobacilli, Lactococci, Streptococci and Leuconostocs, while Bifidobacteria can grow uninhibited.

Application

MUP Selective Supplement is used as a supplement for the preparation of TOS-MUP agar. It is suitable for the enumeration of Bifidobacteria in Dairy products (probiotics, yoghurts, yoghurt like products, and dried milk products such as Bifidobacteria containing infant milk formulae).

Analysis Note

Appearance (clearness): clear
Testing as supplement in TOS-Propionat Agar (1.00043.0500).
Recovery on test medium
- Bifidobacterium animalis subsp. lactis ATCC 27536:
≥ 70 %
- Bifidobacterium breve ATCC 15700: ≥ 70 %
- Bifidobacterium longum ATCC 15707: ≥ 70 %
Growth
- Lactobacillus casei ATCC 393 (WDCM 00100):
no growth
- Lactobacillus delbrueckii subsp. bulgaricus ATCC 11842 (WDCM 00102): no growth
- Streptococcus salivarius subsp. thermophilus DSM 20259: no growth
Incubation: 72 hrs. at 37°C, anaerobic.

Other Notes

Replaces: 1.00045.0017


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library