Key Documents
1.01111
Supelco
Reducer (stainless steel)
for 2" to S40 thread
About This Item
UNSPSC Code:
41121815
NACRES:
NA.03
fitting
for 2" to S40 thread
storage temp.
no temp limit
Related Categories
General description
This reducer connects our barrels and drums with G2 opening to S40-threaded equipment. It is equipped with a G2 outer thread and an S40 outer thread.
from 2" to S40 thread