Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902694

Supelco

Reducer (PE)

for S40 to GL45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

fitting

for S40 to GL45

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Special S40 bottle adapters made of PTFE provide all possible connection options for safe and reliable daily solvents handling. This adapter is equipped with S40 inner thread and GL45 outer thread.
from S40 to GL45

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service