Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.88015

Supelco

Solvent

solvent for volumetric Karl Fischer titration with two component reagents, Aquastar®

Synonym(s):

Karl Fischer Solvent: Karl Fischer two component solvent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601
NACRES:
NA.21

Product Name

Solvent, solvent for volumetric Karl Fischer titration with two component reagents Aquastar®

Quality Level

product line

Aquastar®

form

liquid

reaction suitability

reaction type: volumetric

technique(s)

Karl-Fischer titration: suitable
titration: suitable

transition temp

flash point 11 °C (Methanol)

density

0.90 g/cm3 at 20 °C

format

two component solution

storage temp.

15-25°C

Analysis Note

Application test: passes test

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup - Methanol

Flash Point(C)

11 °C - closed cup - Methanol


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Summary application report for analysis of moisture in D(+)-glucose-monohydrate

Summary application report for analysis of moisture in Ammonia, aqueous

Summary application report for analysis of moisture in D(+)-glucose-monohydrate

Summary application report for analysis of moisture in D(+)-glucose-monohydrate

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service