Skip to Content
MilliporeSigma
All Photos(1)

Documents

1.00543

Supelco

SICAPENT® with indicator

(phosphorus pentoxide drying agent for desiccators) on inert carrier material

Synonym(s):

Phosphorus pentoxide drying agent, granulated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123003
NACRES:
NB.11

Quality Level

form

solid

particle size

0.1-1.6 mm

pH

1.5 (20 °C, 10 g/L in H2O)

bulk density

300 kg/m3

storage temp.

no temp limit

General description

SICAPENT® with indicator is a phosphorus pentoxide drying agent for desiccators. It is also used for drying liquids and filling drying tubes, particularly for fast-flowing gases in drying tubes.

Features and Benefits

The main advantage of using granulated drying agents is the ease of use. Even after significant water uptake (approx. 100% of its own weight), the substance remains in particle form. Hence, subsequent to drying, the agent can be easily removed from the vessel. SICAPENT® drying agents dry well due to their large surface area; it is around 20% faster than standard phosphorus pentoxide.

Linkage

Replaces: SX0137-1

Analysis Note

Water absorption capacity: ≥ 50 %
colour change to blue: conforms

Legal Information

Sicapent is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service