Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.05704

Supelco

Molecular sieve 0.3 nm

beads ~ 2 mm / ~ 10 mesh

Synonym(s):

Molecular sieve 0.3 nm, Sodium aluminium silicate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41123003
EC Index Number:
215-283-8
NACRES:
NB.51
Pricing and availability is not currently available.

Quality Level

form

beads

technique(s)

titration: suitable

surface area

800 m2/g

particle size

~10 mesh
~2 mm

pore size

0.3 nm

pH

8-11 ( in H2O)

mp

>1600 °C

bulk density

700‑750 kg/m3

storage temp.

no temp limit

SMILES string

[Si](=O)=O.[Al](=O)O[Al]=O

InChI

1S/2Al.O2Si.3O/c;;1-3-2;;;

InChI key

HNPSIPDUKPIQMN-UHFFFAOYSA-N

General description

Molecular sieve 0.3 nm is a porous solid that can be used in desiccators and drying tubes to dry gases and absolute solvents. It also changes its color from brown to yellowish at a water uptake of approximately 7-10 g/100 g molecular sieve.

Application

Molecular sieve 0.3 nm can be used as an adsorbent:
  • To remove traces of moisture in chemicals.
  • In ethanol-water mixture purification to obtain pure ethanol.
It can also be used as a catalyst in the condensation of hydroxylamine hydrochloride with aldehydes, ketones, and 1,3-diketones to synthesize aldoximes, ketoximes, and 1,3-dioximes respectively.

Features and Benefits

  • Easy to use
  • Chemically inert
  • Non-toxic
  • Dried liquids can be decanted
  • High adsorption capacity
  • Selective adsorption
  • Eco-friendly

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Does this product come pre-activated? I.e. is it ready to use upon arrival

    1 answer
    1. Yes, Product 1.05704 is a pre-activated or pre-dried and ready-to-use material. The typical water content (reported as a loss on combustion on the lot-specific certificate of analysis) is about 1 percent, versus a water capacity of about 20 percent. Having said that, if there is a need to remove the trace amount of water present, these molecular sieves can be heated to 300–350 °C.

      Helpful?

  4. do you carry zeoliet 13x and MgO variants?

    1 answer
    1. There are 5 product options for the 13X Zeolite. Each offering a different particle size range. See the link below to review:
      https://www.sigmaaldrich.com/substance/molecularsieves13x1234563231696

      There are also a number of Mangesium Oxide product options. See the link below for a list of products:
      https://www.sigmaaldrich.com/substance/magnesiumoxide40301309484

      Helpful?

  5. what is wire mesh size for 3A type 0.3nm ( for support purpose )

    1 answer
    1. This product is sold in a bead format with a mesh size of about 10 nm.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service