Documents
Z682667
Ace drying flask, pear-shaped, with side indents
capacity 1000 mL, joint: ST/NS 24/40, glass
Synonym(s):
Aldrich® drying flask, pear-shaped, with side indents
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
glass
pear-shaped flask
single-neck
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Büchi equivalent, 00424
capacity
1000 mL
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC