Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.00205

Millipore

GranuCult® prime MacConkey (MAC) Agar

according to ISO (21150, 21567, FDA-BAM ), according to EP, according to USP, according to JP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

according to EP
according to ISO (21150, 21567, FDA-BAM )
according to JP
according to USP

Quality Level

form

powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

selective for coliforms: Escherichia spp.
selective for coliforms: Salmonella spp.
selective for coliforms: Shigella spp.

General description

GranuCult® prime MacConkey (MAC) Agar is a selective agar medium that is designed for the isolation and confirmation of Escherichia coli and other Enterobacteriaceae, e.g. Salmonella and Shigella, from the food chain, cosmetic, pharmaceutical and other materials. This culture medium contains crystal violet and bile salts that inhibit the growth of the Gram-positive microbial flora whilst it allows the Gram-negative bacteria to grow. Peptones provide nitrogen, vitamins and amino acids. Lactose and the pH indicator neutral red are used to detect lactose degradation. The fermentation of lactose is causing a local drop down of the pH, resulting in a colour change of the pH indicator neutral red. By this, reddish-pink colonies are formed and the medium around the colonies changes its colour to reddish. Often a bile precipitation zone around the colony is formed. Bile salts and crystal violet inhibits the growth of Gram-positive bacteria. Agar is the solidifying agent.

Application

GranuCult® prime MacConkey (MAC) agar is used to isolate and to confirm Escherichia coli and other Enterobacteriaceae, e.g. Salmonella and Shigella, from the food chain, cosmetic, pharmaceutical and other materials.A variety of bacteria like salmonella spp, shigella spp and coliform bacteria from samples.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service