Skip to Content
MilliporeSigma
All Photos(2)

Documents

09-213

Sigma-Aldrich

Anti-phospho-mTOR (Ser2448) Antibody

Upstate®, from rabbit

Synonym(s):

FRAP1, FRAP, FRAP2, RAFT1, RAPT1, FK506 binding protein 12-rapamycin associated protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pSer2448)

Gene Information

human ... MTOR(2475)

General description

mTOR (Mammalian Target of Rapamycin, aka FRAP, RAPT or RAFT) is a large 289 kDa Ser/Thr protein kinase that regulates cell cycle progression, cell growth, protein synthesis, ribosome biogenesis, and autophagy. mTOR is an evolutionarily conserved member of the Phosphoinositol Kinase-related Kinase (PIKK) family whose activity is regulated by phosphorylation on Ser2448 by Akt in response to insulin or muscle activity. Interestingly, mTOR is the central component of two multimeric kinase complexes consisting of mTOR and numerous other mTOR binding proteins. These two multimeric protein complexes are designated mTORC1 and mTORC2. mTORC1 (mTOR Complex 1) consists of at least mTOR, Raptor, and GβL (mLST8). mTORC1 is known to play a central role in insulin signaling, which is crucial in maintaining metabolic homeostasis. The complex is activated primarily though the PI3 Kinase/Akt pathway. Upon insulin stimulation, Akt activates mTORC1 by phosphorylating and inhibiting TSC (and possibly other yet discovered targets and/or mTOR itself). This inhibits the upstream small GTPase regulator Rheb (Ras homolog enriched in brain). This inhibits the kinase activity of the mTORC1 complex, thus disabling its ability to phosphorylate its downstream targets such as p70 S6K on Thr389 and 4E-BP1 on Thr229. The other mTOR complex, mTORC2 (mTOR Complex 2), is made up of at least mTOR, Rictor, GL, Sin1, Protor 1 and 2. mTORC2 affects cell proliferation and survival primarily by phosphorylating the hydrophobic motif of Akt on Ser473, a well-known effecter of the PI3 Kinase pathway. In addition to phosphorylating Akt, the mTORC2 complex is also known to effect cytoskeletal organization and migration by exerting its effects through Rac, Rho, and PKC. Interestingly, unlike mTORC1, the mTORC2 complex appears to not be inhibited by treatment with rapamycin and for this reason is referred to as the rapamycin-insensitive complex. Defects in both mTOR complexes are associated with a variety of diseases, including cancer and diabetes.

Specificity

Predicted to cross react with rat, mouse, sheep & Rhesus Macaque based on 100% sequence homology.
Recognizes mTOR when phosphorylated on Ser2448

Immunogen

Amino acids encompassing and including phosphorylated Ser2448 of human mTOR

Application

Research Sub Category
PI3K, Akt, & mTOR Signaling

Glucose/Glycogen Metabolism

Insulin/Energy Signaling

Adhesion (CAMs)
This Anti-mTOR Antibody is validated for use in WB for the detection of phospho-mTOR (Ser2448).

Quality

Evaluated by western blot on EGF-treated and untreated A431 cell lysate.

Western Blot Analysis:
This antibody detected mTOR phosphorylated on Ser2448 in EGF-treated and untreated A431 cell lysates.

Target description

~289 kDa observed.
An uncharacterized band may be obsereved at ~59 kDa

Physical form

Purified rabbit polyclonal IgG in storage buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Analysis Note

Control
EGF-treated and untreated A431 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Tateishi et al.
Acta neuropathologica communications, 11(1), 186-186 (2023-11-28)
In IDH-mutant astrocytoma, IDH2 mutation is quite rare and biological mechanisms underlying tumor progression in IDH2-mutant astrocytoma remain elusive. Here, we report a unique case of IDH2 mutant astrocytoma, CNS WHO grade 3 that developed tumor progression. We performed a
Tatsuo Adachi et al.
Drug discoveries & therapeutics, 9(4), 282-288 (2015-04-07)
Some peptides that are highly conserved between insects and mammals have anti-tumor action. Screening for inhibitors of cell growth from animal fluids may provide useful clues to anti-tumor drugs. Inducers of autophagy also have anti-tumor activity. The current authors recently
SWI/SNF chromatin remodeling enzyme ATPases promote cell proliferation in normal mammary epithelial cells.
Cohet, N; Stewart, KM; Mudhasani, R; Asirvatham, AJ; Mallappa, C; Imbalzano, KM; Weaver et al.
Journal of Cellular Physiology null
Yange Liu et al.
BioMed research international, 2017, 9374026-9374026 (2017-04-21)
Antrodia cinnamomea, a folk medicinal mushroom, has numerous biological effects. In this study, we aim to assess whether the antifatigue effects of A. cinnamomea mycelia (AC) and its underlying mechanisms are related to oxidative stress signaling using behavioral mouse models
Giada Poli et al.
Oncotarget, 7(31), 49636-49648 (2016-07-09)
Adrenocortical carcinoma (ACC) is a rare heterogeneous malignancy with poor prognosis. Since radical surgery is the only available treatment, more specific and effective drugs are urgently required. The anti-diabetic drug metformin has been associated with a decreased cancer prevalence and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service