Skip to Content
MilliporeSigma
All Photos(4)

Documents

05-1592

Sigma-Aldrich

Anti-mTOR Antibody, clone 21D8.2

clone 2ID8.2, from mouse

Synonym(s):

FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP-rapamycin associated protein, FKBP12-rapamycin complex-associated prot

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2ID8.2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTOR(2475)

General description

mTOR (Mammalian Target of Rapamycin, aka FRAP, RAPT or RAFT) is a large 289 kDa Ser/Thr protein kinase that regulates cell cycle progression, cell growth, protein synthesis, ribosome biogenesis, and autophagy. mTOR is an evolutionarily conserved member of the Phosphoinositol Kinase-related Kinase (PIKK) family whose activity is regulated by phosphorylation on Ser2448 by Akt in response to insulin or muscle activity. mTORC1 (mTOR Complex 1) consists of at least mTOR, Raptor, and G (mLST8). mTORC1 is known to play a central role in insulin signaling, which is crucial in maintaining metabolic homeostasis. mTORC2 affects cell proliferation and survival primarily by phosphorylating the hydrophobic motif of Akt on Ser473, a well-known effecter of the PI3 Kinase pathway. In addition to phosphorylating Akt, the mTORC2 complex is also known to effect cytoskeletal organization and migration by exerting its effects through Rac, Rho, and PKC. Defects in both mTOR complexes are associated with a variety of diseases, including cancer and diabetes.

Specificity

This antibody recognizes mTOR.

Immunogen

Flag-tagged full-length mTOR protein

Application

Anti-mTOR Antibody, clone 21D8.2 is an antibody against mTOR for use in WB & IC.
Immunocytochemistry Analysis: A 1:500 dilution of this antibody from a previous lot detected mTOR in C2C12, HeLa, and NIH/3T3 cell lysates.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by Western Blot in HEK293 cell lysate.
Western Blot Analysis: 0.5 µg/mL of this antibody detected mTOR on HEK293 cell lysate.

Target description

289 kDa

Linkage

Replaces: 04-385

Physical form

Format: Purified
Protein G Purified
Purified immunoglobulin containing 0.1 M Tris-Glycine, pH 7.4, 150 mM NaCl and 0.05% Sodium Azide.

Storage and Stability

Maintain refrigerated at 2-8°C for 1 year from date of receipt.

Analysis Note

Control
Hek293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sidney Abou Sawan et al.
American journal of physiology. Cell physiology, 315(4), C537-C543 (2018-08-23)
We have recently demonstrated that whole egg ingestion induces a greater muscle protein synthetic (MPS) response when compared with isonitrogenous egg white ingestion after resistance exercise in young men. Our aim was to determine whether whole egg or egg white
Joseph J Bass et al.
Molecular metabolism, 42, 101059-101059 (2020-08-11)
The Vitamin D receptor (VDR) has been positively associated with skeletal muscle mass, function and regeneration. Mechanistic studies have focused on the loss of the receptor, with in vivo whole-body knockout models demonstrating reduced myofibre size and function and impaired muscle
Kuo-Hui Su et al.
Nature cell biology, 18(5), 527-539 (2016-04-05)
To cope with proteotoxic stress, cells attenuate protein synthesis. However, the precise mechanisms underlying this fundamental adaptation remain poorly defined. Here we report that mTORC1 acts as an immediate cellular sensor of proteotoxic stress. Surprisingly, the multifaceted stress-responsive kinase JNK
Somayeh Ahmadiantehrani et al.
Learning & memory (Cold Spring Harbor, N.Y.), 25(6), 273-282 (2018-05-17)
Nonassociative learning is considered simple because it depends on presentation of a single stimulus, but it likely reflects complex molecular signaling. To advance understanding of the molecular mechanisms of one form of nonassociative learning, habituation, for ethologically relevant signals we
Yong-Yan Bi et al.
Molecular medicine reports, 17(1), 1093-1098 (2017-11-09)
Accumulating data strongly suggests that leukocyte immunoglobulin like receptor B1 (PirB) inhibits axonal outgrowth. However, the underlying mechanisms remain unclear. In the present study, cortical neurons of newborn mice were cultured with Nogo‑66 (Nogo‑p4; 4 µmol/l; a PirB ligand) together with

Articles

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Autophagy is a regulated process involved in cell growth, development, and recycling of cytoplasmic components in cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service