Skip to Content
MilliporeSigma
All Photos(2)

Documents

06-1310

Sigma-Aldrich

Anti-JAK2 Antibody

from rabbit

Synonym(s):

Janus kinase 2, Janus kinase 2 (a protein tyrosine kinase), tyrosine-protein kinase JAK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... JAK2(3717)
mouse ... Jak2(16452)
rat ... Jak2(24514)

Related Categories

General description

The Janus family of tyrosine kinases (JAK) includes JAK1, JAK2, JAK3 and Tyk2. This family associates with a variety of cytokine receptors and is activated by ligand binding. Upon ligand binding to cytokine receptors, JAKs phosphorylate themselves and their associated receptors. This provides multiple binding sites for signaling proteins containing SH2 or other phospho-tyrosine-binding domains, including STATs, Shc, insulin receptor substrates and FAK.

Specificity

This antibody recognizes the protein kinase 1 domain of JAK2.

Immunogen

Epitope: Protein kinase 1 domain
KLH-conjugated linear peptide corresponding to the protein kinase 1 domain of mouse JAK2.

Application

Research Category
Inflammation & Immunology

Signaling
Research Sub Category
Immunological Signaling

Kinases & Phosphatases
This Anti-JAK2 Antibody is validated for use in WB, IP for the detection of JAK2.
Western Blot Analysis: 1 µg/mL from a previous lot detected JAK2 in 10 µg of L6 cell lysate.

Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated JAK2 from RAW264.7 cell lysate.

Quality

Evaluated by Western Blot in RAW264.7 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected JAK2 in 10 µg of RAW264.7 cell lysate.

Target description

~130 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
RAW264.7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiahai Shi et al.
Experimental hematology, 44(11), 1044-1058 (2016-10-27)
JAK2 V617F is a mutant-activated JAK2 kinase found in most polycythemia vera (PV) patients; it skews normal proliferation and differentiation of hematopoietic stem and progenitor cells and simulates aberrant expansion of erythroid progenitors. JAK2 V617F is known to activate some
Sandrine Medves et al.
Haematologica, 96(10), 1406-1414 (2011-06-21)
KANK1-PDGFRB is a fusion gene generated by the t(5;9) translocation between KANK1 and the platelet-derived growth factor receptor beta gene PDGFRB. This hybrid was identified in a myeloproliferative neoplasm featuring severe thrombocythemia, in the absence of the JAK2 V617F mutation.
Xiaomei Li et al.
Virology, 551, 75-83 (2020-08-25)
Porcine epidemic diarrhea (PED) virus (PEDV) is a coronavirus that primarily infects porcine intestinal epithelial cells and causes severe diarrhea and high fatality in piglets. A77 1726 is the active metabolite of leflunomide, a clinically approved anti-rheumatoid arthritis (RA) drug.
Alice H Grant et al.
International journal of molecular sciences, 24(7) (2023-04-14)
Overactive Janus kinases (JAKs) are known to drive leukemia, making them well-suited targets for treatment. We sought to identify new JAK-activating mutations and instead found a JAK1-inactivating pseudokinase mutation, V666G. In contrast to other pseudokinase mutations that canonically lead to
Ting Wang et al.
EMBO molecular medicine, 9(8), 1150-1164 (2017-06-03)
The endocrine system is crucial for maintaining whole-body homeostasis. Little is known regarding endocrine hormones secreted by the heart other than atrial/brain natriuretic peptides discovered over 30 years ago. Here, we identify growth differentiation factor 15 (GDF15) as a heart-derived hormone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service