Skip to Content
MilliporeSigma
All Photos(2)

Documents

SAB4501599

Sigma-Aldrich

Anti-JAK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

JAK-2, JAK2, Janus kinase 2, kinase Jak2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... JAK2(3717)

General description

Anti-JAK2 antibody detects endogenous levels of total JAK2 protein.
JAK2 (janus kinase 2) encodes a tyrosine kinase. It belongs to the the Jak family of proteins. It is located on human chromosome 9p24.

Immunogen

The antiserum was produced against synthesized peptide derived from human JAK2.

Immunogen Range: 541-590

Biochem/physiol Actions

JAK2 (janus kinase 2) plays an important role in the activities of cytokines receptors. Overexpression of JAK2 results in neuroblastoma. JAK2 mutations promote primary resistance to PD-1 (anti-programmed death protein 1) blockade therapy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cancer-Associated Fibroblasts Share Characteristics and Protumorigenic Activity with Mesenchymal Stromal Cells
Borriello L, et al.
Cancer Research (2017)
Xiaoping Pan et al.
Life sciences, 214, 124-135 (2018-11-06)
Although the anti-neoplastic effects of physcion are well documented, its specific action in hepatocellular carcinoma (HCC) is not understood. Taken together, physcion is a promising drug candidate for the treatment and prevention of HCC. Cell Counting Kit-8 (CCK-8) assay utilized
Jak2 is essential for signaling through a variety of cytokine receptors
Parganas E, et al.
Cell, 93(3), 385-395 (1998)
Hodgkin's lymphoma cell lines are characterized by frequent aberrations on chromosomes 2p and 9p including REL and JAK2
Joos S, et al.
International Journal of Cancer. Journal International Du Cancer, 103(4), 489-495 (2003)
Primary Resistance to PD-1 Blockade Mediated by JAK1/2 Mutations.
Shin DS
Cancer Discovery, 7(2), 188-201 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service