Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09-241

Sigma-Aldrich

Anti-phospho JAK-2 (Tyr570) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Janus kinase 2, Janus kinase 2 (a protein tyrosine kinase), tyrosine-protein kinase JAK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, pig, horse

species reactivity (predicted by homology)

primate (based on 100% sequence homology), equine (based on 100% sequence homology), porcine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr570)

Gene Information

human ... JAK2(3717)

General description

JAK-2 (Janus Kinase 2) belongs to the emerging family of non-receptor Janus tyrosine kinases, which regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK-2. JAK-2 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to CIS elements upstream of target gene promoters to regulate transcription. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival and host resistance to pathogens. Altering JAK/Stat signaling to reduce cytokine induced pro-inflammatory responses represents an attractive target for anti-inflammatory therapies. Within the JAK-2 kinase domain, there is a region that has considerable sequence homology to the regulatory region of the insulin receptor.

Specificity

This antibody recognizes JAK-2 when phosphorylated on Tyr570.

Immunogen

Epitope: Phosphorylated Tyr570
KLH-conjugated linear peptide corresponding to human JAK-2 phosphorylated at Tyr570.

Application

Research Category
Signaling
Research Sub Category
Cytokines & Cytokine Receptors
This Anti-phospho JAK-2 (Tyr570) Antibody is validated for use in WB, WB, WB for the detection of phospho JAK-2 (Tyr570).

Quality

Evaluated by Western Blot in transfected HEK293T cell lysates.

Western Blot Analysis: 0.05 ug/mL of this antibody detected JAK-2 on 10 µg of transfected HEK293T cell lysates.

Target description

~ 125 kDa

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Transfected HEK293T cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Probiotics down-regulate genes in Salmonella enterica serovar typhimurium pathogenicity islands 1 and 2.
Mohamed A Bayoumi,Mansel W Griffiths
Journal of Food Protection null
Rita Andraos et al.
Cancer discovery, 2(6), 512-523 (2012-06-12)
Janus kinase (JAK) inhibitors are being developed for the treatment of rheumatoid arthritis, psoriasis, myeloproliferative neoplasms, and leukemias. Most of these drugs target the ATP-binding pocket and stabilize the active conformation of the JAK kinases. This type I binding mode
Michael Xiang et al.
Blood, 128(14), 1845-1853 (2016-08-18)
The oncogenic transcription factor signal transducer and activator of transcription 3 (STAT3) is frequently activated inappropriately in a wide range of hematological and solid cancers, but clinically available therapies targeting STAT3 are lacking. Using a computational strategy to identify compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service