Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04-855

Sigma-Aldrich

Anti-GluR1 Antibody, clone C3T, rabbit monoclonal

culture supernatant, clone C3T, Upstate®

Synonym(s):

Anti-GluA1, Anti-GluR-1, Anti-GluR-A, Anti-GluR-K1, Anti-GluR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

C3T, monoclonal

species reactivity

human, chimpanzee, rat, mouse

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIA1(2890)

General description

L glutamate is the major excitatory neurotransmitter in the central nervous system. Glutamatergic neurotransmission is involved in most aspects of normal brain function and can be perturbed in many neuropathologic conditions. The metabotropic glutamate receptors are a family of G protein coupled receptors that have been divided into 3 groups. Group I includes GRM1 and these receptors have been shown to activate phospholipase C. GRM1 expression has been reported in various regions of the brain and spinal cord. ESTs have been isolated from brain and eye libraries.

Specificity

Predicted to cross-react with Chimpanzee based pm 100% sequence homology.
Recognizes GluR1, Mr 106 kDa.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 858-868 (..TLPRNSGAGAS..) of human GluR1 (amino acids 840-850 of mature human GluR1)

Application

Anti-GluR1 Antibody, clone C3T detects level of GluR1 & has been published & validated for use in IH, IP & WB.
Immunoprecipitation: 2 μL of a previous lot immunoprecipitated GluR1 from rat brain microsomal preparation.

Immunohistochemistry: A previous lot of this antibody has been shown to detect GluR1 in human, mouse and rat tissues.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Routinely evaluated by Western blotting of rat brain microsomal preparation.

Western Blotting Analysis (WB): A 1:5,000-1:10,000 dilution of this lot detected GluR1 in rat brain microsomal preparation (Catalog # 12-144).

Target description

106 kDa

Linkage

Replaces: 05-855

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Routinely evaluated by immunoblot on rat brain microsomal preparation (12-144)

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A L Mammen et al.
The Journal of biological chemistry, 272(51), 32528-32533 (1998-01-24)
Modulation of alpha-amino-3-hydroxy-5-methylisoxazole-4-propionic Acid (AMPA) receptors in the brain by protein phosphorylation may play a crucial role in the regulation of synaptic plasticity. Previous studies have demonstrated that calmodulin (CaM) kinase II can phosphorylate and modulate AMPA receptors. However, the
Gonadectomy differentially regulates steroid receptor coactivator-1 and synaptic proteins in the hippocampus of adult female and male C57BL/6 mice.
Chen Bian,Kongjiang Zhu,Li Yang,Sen Lin,Shurong Li,Bingyin Su,Jiqiang Zhang
Synapse null
Linli Qiu et al.
The Journal of steroid biochemistry and molecular biology, 156, 23-31 (2015-11-27)
Androgens have been proposed to play important roles in the regulation of hippocampus function either directly, through the androgen receptor (AR), or indirectly, through estrogen receptors (ERs), after aromatization into estradiol. Steroid receptor coactivator-1 (SRC-1) is present in the hippocampus
Zhexing Wen et al.
Nature, 515(7527), 414-418 (2014-08-19)
Dysregulated neurodevelopment with altered structural and functional connectivity is believed to underlie many neuropsychiatric disorders, and 'a disease of synapses' is the major hypothesis for the biological basis of schizophrenia. Although this hypothesis has gained indirect support from human post-mortem
Cellular localizations of AMPA glutamate receptors within the basal forebrain magnocellular complex of rat and monkey.
Martin, L J, et al.
The Journal of Neuroscience, 13, 2249-2263 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service