Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05-1540-M

Sigma-Aldrich

Anti-Gemin2 Antibody, clone 2E17

clone 2E17, from mouse

Synonym(s):

GEMIN2, Gemin2, Component of gems, SMN interacting protein 1-delta, SMN-interacting protein 1, survival interacting protein 1, survival of motor neuron protein interacting protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2E17, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... GEMIN2(8487)

General description

Gemin-2 (also known as SIP1 or SMN interacting protein1) is a part of the SMN complex. The SMN complex plays a role in RNA metabolism and in splicing regulation. As part of the SMN complex Gemin-2 interacts directly with Gemin-5 in a heteromeric complex that includes Gemin-3, Gemin-4, Gemin-6. This macromolecular complex (SMN complex) mediates assembly of spliceosomal snRNA complexes containing U1 and U5.

Specificity

Antibody recognizes Gemin-2.

Immunogen

His-tagged recombinant protein corresponding to human Gemin-2.

Application

Anti-Gemin2 Antibody, clone 2E17 is a Mouse Monoclonal Antibody for detection of Gemin2 also known as SMN interacting protein 1-delta & has been validated in WB, ICC, ELISA & IP.
Immunocytochemistry:
Cited in Battle, D.J., et. al (2007). J. Biol. Chem. 282: 27953 - 27959 and Zhang, Z., et. al (2008). Cell, 133:85-600. using a representative lot.
ELISA:
Cited in Zhang, Z., et. al (2008). Cell, 133:85-600.
Immunoprecipitation:
Cited in Battle, D.J., et. al(2007). J. Biol. Chem. 282: 27953 - 27959 using a representative lot.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in HeLa cell lysates.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Gemin-2 in HeLa cell lysates.

Target description

32 kDa Calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse IgG1κ supernatant in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service