Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

V-036

Supelco

(±)-Voriconazole-D3 solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Voriconazole-D₃ Isomer Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11D3F3N5O
Molecular Weight:
352.33
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

A stable-labeled internal standard suitable for quantification of voriconazole levels in blood, serum, or plasma by HPLC or LC-MS/MS for applications such as therapeutic drug monitoring or clinical and diagnostic testing. Voriconazole, sold under the trade name Vfend®, is a triazole antifungal drug prescribed for treatment of serious, invasive fungal infections.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Vfend is a registered trademark of Pfizer, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service