Skip to Content
MilliporeSigma

Skip To

K-004

Ketoconazole solution

2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H28Cl2N4O4
CAS Number:
Molecular Weight:
531.43
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

2.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=C(C=CC(Cl)=C1)[C@@]2(O[C@@H](COC3=CC=C(N4CCN(C(C)=O)CC4)C=C3)CO2)CN5C=NC=C5

InChI

1S/C26H28Cl2N4O4/c1-19(33)31-10-12-32(13-11-31)21-3-5-22(6-4-21)34-15-23-16-35-26(36-23,17-30-9-8-29-18-30)24-7-2-20(27)14-25(24)28/h2-9,14,18,23H,10-13,15-17H2,1H3/t23-,26-/m1/s1

InChI key

XMAYWYJOQHXEEK-ZEQKJWHPSA-N

General description

Ketoconazole, sold under trade names such as Nizoral®, Sebizole, Ketomed, and Fungarest, is an antifungal drug used in the treatment and prevention of superficial and systemic fungal infections. Ketoconazole is routinely assigned therapeutic drug monitoring due to its high toxicity and narrow therapeutic range. This Certified Spiking Solution® is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of ketoconazole in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Nizoral is a registered trademark of Johnson & Johnson
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1385K06000001356508
grade

certified reference material

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

single component solution

format

neat

format

neat

format

neat

manufacturer/tradename

Cerilliant®

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

USP

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

storage temp.

−20°C

storage temp.

2-30°C

storage temp.

-

storage temp.

-

concentration

2.0 mg/mL in methanol

concentration

-

concentration

-

concentration

-


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters