Skip to Content
MilliporeSigma
All Photos(1)

Documents

U-009

Supelco

U-49900 solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26Cl2N2O
CAS Number:
Molecular Weight:
357.32
UNSPSC Code:
41116107

grade

certified reference material

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

U-49900 is a highly potent opioid analgesic that has been confirmed in many opioid overdoses. This Certified Spiking Solution® is suitable as starting material for use in calibrators or controls for a variety of LC/MS or GC/MS applications from clinical toxicology, urine drug testing or forensic analysis applications.

This Cerilliant product is part of the Centers for Disease Control and Prevention (CDC) Opioid CRM Kit including more than 40 CRMs to be used as part of CDC′s laboratory response to the opioid epidemic. The kits are provided free of charge to drug testing laboratories located in the United States. A requesting lab must have current DEA registration and must comply with all respective state and local regulations. Visit www.cerilliant.com for instructions on requesting a free Opioid CRM Kit. Please note, CDC will review and approve all order requests prior to shipment.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service