Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BM-1RYTJ1

Millipore

Auto2D® Tricine Type Reagent Kit

reagent kit for the Auto2D® 2-D Electrophoresis Device, for better resolution on low molecular weight range

Synonym(s):

2D Electrophoresis, 2DE, 2DE Reagent, Tricine, Tris-Tricine, Tris-Tricine Reagent Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

product line

Auto2D®

technique(s)

SDS-PAGE: suitable
western blot: suitable

storage temp.

2-8°C

General description

Tricine Type Reagent Kit is used for the Auto2D® 2-D Electrophoresis Device with better resolution on low molecular weight range than Glycine Type Reagent Kit.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Components

  • Rehydration Solution Kit
  • DTT Kit
  • Tricine Type Equilibration Buffer
  • Anode + Buffer
  • Tricine Type Cathode - Buffer

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Protocols

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

Related Content

Auto2D® 2-D Gel Electrophoresis Device frequently asked questions for 2-D PAGE and additional 2DE protein separation workflow needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service