Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N-115

Supelco

Naloxone-D5 solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16NO4D5
CAS Number:
Molecular Weight:
332.41
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[2H]\C([2H])=C(/[2H])C([2H])([2H])N1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@]2(O)CCC4=O)c35

InChI

1S/C19H21NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,14,17,21,23H,1,5-10H2/t14-,17+,18+,19-/m1/s1/i1D2,2D,8D2

InChI key

UZHSEJADLWPNLE-HEEDNVRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for numerous quantitative LC/MS and GC/MS applications in forensic analysis, clinical toxicology testing, pharmaceutical analysis and isotope dilution methods. Naloxone is an opiate drug used to counter the effects of opioid overdose.

Application


  • Naloxone-D₅ analytical standard: Naloxone-D₅ is a stable isotopic label extensively utilized as an analytical standard in mass spectrometry to study the pharmacokinetics of naloxone. This application is critical for understanding naloxone′s metabolism and clearance rates in opioid overdose scenarios, thereby enhancing therapeutic strategies and drug safety (Xu et al., 2018).


Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service