Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L-034

Supelco

Levorphanol-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20D3NO
CAS Number:
Molecular Weight:
260.39
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC([C@@]2(CCCC3)[C@]3([H])[C@H](N(C([2H])([2H])[2H])CC2)C4)=C4C=C1

InChI

1S/C17H23NO/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17/h5-6,11,14,16,19H,2-4,7-10H2,1H3/t14-,16+,17+/m0/s1/i1D3

InChI key

JAQUASYNZVUNQP-ZHFAENJZSA-N

General description

A Certified Spiking Solution® suitable for use in clinical toxicology, forensic analysis, urine drug testing, or prescription monitoring as an internal standard to quantitate levorphanol levels in urine or blood or for isotope dilution methods by LC-MS/MS or GC/MS. Levorphanol is an opiate drug sold under the trade name Levo-Dromoran to treat severe pain.

Application


  • Forensic toxicology: Levorphanol-D₃ was employed as an internal standard in the development of an enantioselective high-performance liquid chromatography-tandem mass spectrometry method. This method quantitatively determines methorphan and its O-demethylated metabolite in human blood, with applications to post-mortem samples. This study underscores the significance of stable isotope-labeled Levorphanol in enhancing the precision and reliability of forensic toxicology analyses (Lo Faro et al., 2023).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service