Skip to Content
MilliporeSigma
All Photos(1)

Documents

N-103

Supelco

6α-Naloxol solution

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO4
CAS Number:
Molecular Weight:
329.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=C(O2)C([C@]([C@@H]2[C@@H](O)CC3)(CCN4CC=C)[C@]3(O)[C@H]4C5)=C5C=C1

InChI

1S/C19H23NO4/c1-2-8-20-9-7-18-15-11-3-4-12(21)16(15)24-17(18)13(22)5-6-19(18,23)14(20)10-11/h2-4,13-14,17,21-23H,1,5-10H2/t13-,14+,17-,18-,19+/m0/s1

InChI key

HMWHERQFMBEHNG-AQQQZIQISA-N

General description

6α-Naloxol is a metabolite of naloxone, an opiate drug used to counter the effects of opioid overdose. This certified reference solution is suitable as starting material for preparation of calibrators and controls in naloxone testing by LC/MS or GC/MS for forensic analysis, urine drug testing, pharmaceutical research or clinical toxicology applications. In the US, naloxone has become more widely accessible to the general public as a result of state, federal and local initiatives developed to decrease heroin overdose deaths.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Todd A Verner et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 287(2), R411-R421 (2004-03-20)
The aim of this study was to examine the cardiorespiratory effects of chemically stimulating neurons in the midline medulla oblongata (MM) of artificially ventilated and freely breathing anesthetized rats. Earlier studies reported that stimulation of the MM elicits increases or
Daniel Erskine et al.
Movement disorders : official journal of the Movement Disorder Society, 33(6), 982-991 (2018-03-24)
Dementia with Lewy bodies is characterized by transient clinical features, including fluctuating cognition and visual hallucinations, implicating dysfunction of cerebral hub regions, such as the pulvinar nuclei of the thalamus. However, the pulvinar is typically only mildly affected by Lewy
Abdul Bari Muhammad et al.
American journal of physiology. Endocrinology and metabolism, 313(2), E148-E166 (2017-03-09)
Menin is a scaffold protein that interacts with several epigenetic mediators to regulate gene transcription, and suppresses pancreatic β-cell proliferation. Tamoxifen-inducible deletion of multiple endocrine neoplasia type 1 (MEN1) gene, which encodes the protein menin, increases β-cell mass in multiple
Amila Suraweera et al.
Communications biology, 4(1), 484-484 (2021-04-21)
Genomic stability is critical for normal cellular function and its deregulation is a universal hallmark of cancer. Here we outline a previously undescribed role of COMMD4 in maintaining genomic stability, by regulation of chromatin remodelling at sites of DNA double-strand
Sebastian Werneburg et al.
Immunity, 52(1), 167-182 (2019-12-31)
Multiple sclerosis (MS) is a demyelinating, autoimmune disease of the central nervous system. While work has focused on myelin and axon loss in MS, less is known about mechanisms underlying synaptic changes. Using postmortem human MS tissue, a preclinical nonhuman

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service