Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H-134

Supelco

Hydroxynorketamine hydrochloride solution

1.0 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14ClNO2 · HCl
Molecular Weight:
276.16
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

Hydroxynorketamine (HNK), or 6-hydroxynorketamine, is a secondary metabolite of the anesthetic, dissociative, and antidepressant drug ketamine. It is formed by hydroxylation of the intermediate norketamine. This certified Snap-N-Spike® solution is suitable for numerous GC/MS or LC/MS applications in forensic analysis, urine drug testing, clinical toxicology, and clinical or diagnostic testing. Ketamine, sold as Ketanest, Ketaset and Ketalar, is an NMDA receptor antagonist used for the induction and maintenance of general anesthesia in human and veterinary medicine. The drug is often abused at clubs, raves, or parties for its noted hallucingenic and dissociative properties and prescribed by some doctors as an off-label treatment for depression.

Application

  • Hydroxynorketamine HCl for neuropharmacological research: Hydroxynorketamine hydrochloride, a novel NMDA receptor modulator, has shown potential in enhancing synaptic plasticity. This compound provides a promising avenue for exploring neuroprotective strategies in neuropharmacological studies (Fountoulakis et al., 2023).
  • Antidepressant effects of Hydroxynorketamine HCl in preclinical studies: Studies have demonstrated that Hydroxynorketamine hydrochloride exhibits significant antidepressant effects, making it a valuable candidate for treatment-resistant depression research. This compound′s unique mechanism involving the NMDA receptor highlights its potential in psychiatric treatments (Gilbert et al., 2021).
  • Synaptic plasticity research using Hydroxynorketamine hydrochloride: Hydroxynorketamine hydrochloride′s role in modulating synaptic plasticity has been studied, emphasizing its therapeutic potential in neurodegenerative diseases and cognitive disorders. This research underlines the importance of Hydroxynorketamine as a tool in understanding the complexities of synaptic functions (Evans et al., 2018).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service