Skip to Content
MilliporeSigma

SML1875

(2S,6S)-Hydroxynorketamine hydrochloride

≥98% (HPLC), Ketamine metabolite, powder

Synonym(s):

(2S,6S)-2-Amino-2-(2-chlorophenyl)-6-hydroxy-cyclohexanone hydrochloride, (2S,6S)-HNK hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H14ClNO2 · HCl
CAS Number:
Molecular Weight:
276.16
UNSPSC Code:
12352116
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

(2S,6S)-Hydroxynorketamine hydrochloride, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +95 to +115°, c = 1 in H2O

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada

storage condition

desiccated

color

white to beige

solubility

H2O: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

[H]Cl.ClC1=CC=CC=C1[C@](CCC[C@@H]2O)(N)C2=O

InChI

1S/C12H14ClNO2.ClH/c13-9-5-2-1-4-8(9)12(14)7-3-6-10(15)11(12)16;/h1-2,4-5,10,15H,3,6-7,14H2;1H/t10-,12-;/m0./s1

InChI key

ZQJVHBJDLMIKJK-JGAZGGJJSA-N

Biochem/physiol Actions

(2S,6S)-Hydroxynorketamine hydrochlorideKetamine is a metabolite associated with increased locomotor activity and motor incoordination.
(2S,6S)-Hydroxynorketamine hydrochlorideKetamine is a metabolite associated with increased locomotor activity and motor incoordination.
It has been found that the NMDAR antagonist (R,S)-ketamine must be metabolized to (2S,6S;2R,6R)-hydroxynorketamine (HNK) to have antidepressant effects. The (2R,6R)-HNK enantiomer appears to be the enantiomer most responsible for antidepressant effects, while (2S,6S)-hydroxynorketamine was associated with increased locomotor activity and motor incoordination. (2S,6S)-HNK, was also found to increase the function of the mammalian target of rapamycin (mTOR) 2-fold at 0.05 nM. (2S,6S)-HNK and (2R,6R)-HNK both inhibited intracellular concentrations of D-serine, an endogenous NMDA receptor co-agonist, with IC50 values of 0.18 nM and 0.68 nM, respectively. Neither bind with high affinity to NMDA receptors, with Ki values of 21.19 μM and > 100 μM for (2S,6S)-HNK and (2R,6R)-HNK, respectively.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library