Skip to Content
MilliporeSigma
All Photos(1)

Documents

F-038

Supelco

Fluoxetine-D6 oxalate solution

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17D6H12F3NO · C2H2O4
Molecular Weight:
405.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C(C(OC2=CC=C(C(F)(F)F)C=C2)([2H])CCNC)C([2H])=C([2H])C([2H])=C1[2H].O=C(O)C(O)=O

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® suitable as an internal standard for quantitation of Fluoxetine levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. Fluoxetine is a pharmaceutical antidepressant which functions as a selective serotonin reuptake inhibitor (SSRI). Fluoxetine is sold under the trade name Prozac for treatment of psychological conditions including major depression and obsessive-compulsive disorder.

Application

A Certified Spiking Solution® suitable as an internal standard for quantitation of Fluoxetine levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. Fluoxetine is a pharmaceutical antidepressant which functions as a selective serotonin reuptake inhibitor (SSRI). Fluoxetine is sold under the trade name Prozac® for treatment of psychological conditions including major depression and obsessive-compulsive disorder.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Prozac is a registered trademark of Eli Lilly and Co.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service