Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L-911

Supelco

Lorazepam-d4 solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D4H6Cl2N2O2
CAS Number:
Molecular Weight:
325.18
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

ClC1=CC=C2C(C(C3=C([2H])C([2H])=C([2H])C([2H])=C3Cl)=NC(O)C(N2)=O)=C1

InChI

1S/C15H10Cl2N2O2/c16-8-5-6-12-10(7-8)13(19-15(21)14(20)18-12)9-3-1-2-4-11(9)17/h1-7,15,21H,(H,18,20)/i1D,2D,3D,4D

InChI key

DIWRORZWFLOCLC-RHQRLBAQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard suitable for quantitation of lorazepam levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, or forensic analysis. Lorazepam, also known by trade names Ativan® or Orfidal, is a benzodiazepine often prescribed for treatment of anxiety disorders.

Application



  • Application in toxicological analysis: Lorazepam-d₄ was utilized as an analytical reference standard in a study aimed at the detection and quantification of lorazepam in human hair by GC-MS/NCI, assisting in forensic investigations related to traffic accidents (Cirimele et al., 1996).


  • Confirmation of urinary benzodiazepine metabolites: A research paper detailed the use of Lorazepam-d₄ in GC-MS methodologies to confirm the presence of benzodiazepine metabolites in urine, providing crucial data for both clinical and forensic settings (Meatherall, 1994).


Legal Information

Ativan is a registered trademark of American Home Product Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service